Pelargonic acid ≥96%

Danger

Synonyms: Nonanoic acid, Pelargonic Acid

N29902-1L N29902-500ML
220200-153EA 74.52 USD
220200-153 220200-154
Pelargonic acid ≥96%
Pelargonic acid
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 12 °C
Density: 0.9055 g/cm³ (20 °C)
Flash Pt: 133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004433
CAS Number: 112-05-0
EINECS: 203-931-2
UN: 3265
ADR: 8,III
Merck Index: 12,07198

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR