1-Naphthyl boronic acid

Warning

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

N257-25G N257-5G
220199-371EA 24.71 USD
220199-371 220200-066
1-Naphthyl boronic acid
1-Naphthyl boronic acid
Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
Melting Pt: 259 °C
MDL Number: MFCD00019722
CAS Number: 13922-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR