3-Nitro-o-phenylenediamine ≥98%

Synonyms: 3-Nitrobenzene-1,2-diamine, 3-Nitro-1,2-phenylenediamine

N21308-5G N21308-1G
220200-047EA 184.62 USD
220200-047 220200-046
3-Nitro-o-phenylenediamine ≥98%
3-Nitro-o-phenylenediamine
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 158…159 °C
MDL Number: MFCD00007722
CAS Number: 3694-52-8
EINECS: 223-013-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR