5-Nitroisatin

Synonyms: 5-Nitro-1H-indole-2,3-dione, 5-Nitroindoline-2,3-dione

N17807-25G N17807-5G
220196-831EA 329.54 USD
220196-831 220196-832
5-Nitroisatin
5-Nitroisatin
Formula: C₈H₄N₂O₄
MW: 192.13 g/mol
Melting Pt: 255…258 °C
MDL Number: MFCD00005720
CAS Number: 611-09-6
EINECS: 210-252-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR