Neocuproine hydrochloride hydrate, crystalline, Bioreagent reagent for copper

Warning

Synonyms: 2,9-Dimethyl-1,10-phenanthroline hydrochloride hydrate

N1626-5G N1626-25G N1626-100G N1626-10G
220199-624EA 197.31 USD
220199-624 220199-623 220199-621 220199-622
Neocuproine hydrochloride hydrate, crystalline, Bioreagent reagent for copper
Neocuproine hydrochloride hydrate

Suitable for determination of copper.

Formula: C₁₄H₁₂N₂·HCl·xH₂O
MW: 244.72 g/mol (anhy)
Melting Pt: 250 °C (dec., lit.)
MDL Number: MFCD00150062
CAS Number: 332360-00-6
EINECS: 230-732-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR