1,8-Naphthalenedicarboxylic anhydride

Warning

Synonyms: Naphthalene-1,8-dicarboxylic anhydride, Benzo[de]isochromene-1,3-dione, 1,8-Naphthalic anhydride

N1607-500G N1607-100G
220199-616EA 163.46 USD
220199-616 220199-615
1,8-Naphthalenedicarboxylic anhydride
1,8-Naphthalenedicarboxylic anhydride
Formula: C₁₂H₆O₃
MW: 198.18 g/mol
Boiling Pt: 425 °C (1013 hPa)
Melting Pt: 267…270 °C
MDL Number: MFCD00006925
CAS Number: 81-84-5
EINECS: 201-380-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR