β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate ≥99%

Synonyms: Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD

N1511-5G N1511-1G N1511-250MG
220199-839EA 796.92 USD
220199-839 220199-837 220199-838
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate ≥99%
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate
Formula: C₂₁H₂₇N₇O₁₄P₂·H₂O
Melting Pt: 140 to 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036253
CAS Number: 53-84-9
EINECS: 200-184-4
Merck Index: 13,06370

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR