Neocuproine

Synonyms: 2,9-Dimethyl-1,10-phenanthroline

N1501-25G N1501-5G N1501-1G
220199-769EA 282.15 USD
220199-769 220199-770 220199-351
Neocuproine
Neocuproine
Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00004973
CAS Number: 484-11-7
EINECS: 207-601-9
Merck Index: 13,06477

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR