α-Bromo-4-nitrotoluene ≥99%

Danger

Synonyms: 4-Nitrobenzyl bromide, 1-(Bromomethyl)-4-nitrobenzene, α-Bromo-p-nitrotoluene

N13054-25G N13054-100G
220198-457EA 49.22 USD
220198-457 220198-456
α-Bromo-4-nitrotoluene ≥99%
α-Bromo-4-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 94 - 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007373
CAS Number: 100-11-8
EINECS: 202-820-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR