4-Nitrobenzyl alcohol ≥99%

Synonyms: (4-Nitrophenyl)methanol

N12821-100G N12821-25G
220199-760EA 136.77 USD
220199-760 220199-761
4-Nitrobenzyl alcohol ≥99%
4-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 90…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007376
CAS Number: 619-73-8
EINECS: 210-611-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR