2-Nitrobenzyl alcohol ≥97%

Synonyms: (2-Nitrophenyl)methanol

N12805-25G N12805-100G
220199-759EA 186.31 USD
220199-759 220199-758
2-Nitrobenzyl alcohol ≥97%
2-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 71…72 °C
MDL Number: MFCD00007186
CAS Number: 612-25-9
EINECS: 210-302-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR