2-Nitrobenzaldehyde ≥98%

N10802-100G N10802-25G N10802-2G
220198-589EA 138.92 USD
220198-589 220198-590 220198-591
2-Nitrobenzaldehyde ≥98%
2-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 41…43 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Merck Index: 12,06684

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR