Methoxyacetyl chloride ≥97%

M9653-1G M9653-10G
220199-427EA 48.46 USD
220199-427 220199-426
Methoxyacetyl chloride ≥97%
Methoxyacetyl chloride
Formula: C₃H₅ClO₂
MW: 108.52 g/mol
MDL Number: MFCD00000728
CAS Number: 38870-89-2
EINECS: 254-169-2
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR