DL-Methionine ≥99%

Synonyms: 2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid

M9500-1KG M9500-100G M9500-500G
220197-822EA 173.54 USD
220197-822 220197-821 220197-823
DL-Methionine ≥99%
DL-Methionine
Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Melting Pt: ∼136 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063096
CAS Number: 59-51-8
EINECS: 200-432-1
Merck Index: 13,06004

Order Now

Specification Test Results

Assay 99% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR