3'-Methoxyacetophenone ≥97%

Synonyms: 3′-Methoxyacetophenone

M9408-100G M9408-25G
220199-382EA 180.92 USD
220199-382 220199-383
3'-Methoxyacetophenone ≥97%
3'-Methoxyacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008736
CAS Number: 586-37-8
EINECS: 209-573-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR