D-Methionine ≥98%

Synonyms: D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH

M9375-5G M9375-25G
220199-325EA 63.63 USD
220199-325 220199-324
D-Methionine ≥98%
D-Methionine
Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 136 °C
MDL Number: MFCD00002622
CAS Number: 348-67-4
EINECS: 206-483-6
Merck Index: 13,06004

Order Now

Specification Test Results

Assay 98% TLC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR