2'-Methoxyacetophenone ≥99%

Synonyms: 1-(2-Methoxyphenyl)ethanone

M9203-25G M9203-100G
220199-310EA 117.38 USD
220199-310 220199-309
2'-Methoxyacetophenone ≥99%
2'-Methoxyacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 131 °C (1013 hPa)
Density: 1.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008725
CAS Number: 579-74-8
EINECS: 209-446-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR