Malachite green oxalate, certified by the Biological Stain Commission
|
Danger
|
Synonyms:
Malachite green hemioxalate, N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate, Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalate, Basic Green 4, N-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)
M9015-100G
M9015-25G
220199-255EA
254.8
USD
220199-255
220199-256
Malachite green oxalate, certified by the Biological Stain Commission
Malachite green oxalate
Formula:
C₅₂H₅₄N₄O₁₂ MW: 927.02 g/mol Melting Pt: ~ 159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011766 CAS Number: 2437-29-8 EINECS: 219-441-7 UN: 2811 ADR: 6.1,III Merck Index: 13,05722 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...