β-(N-Morpholino)ethanesufonic acid (MES) monohydrate

Synonyms: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate

M8250-25KG M8250-250G M8250-1KG M8250-25G M8250-100G
220198-997EA 12446.15 USD
220198-997 220198-995 220198-994 220198-996 220198-993
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
Melting Pt: ? 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Order Now

Specification Test Results

Assay 99.5% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR