N-Methyl-N-(trimethylsilyl)trifluoroacetamide

Danger

Synonyms: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide

M7891-5G M7891-25G
220198-885EA 124.92 USD
220198-885 220198-884
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Formula: C₆H₁₂F₃NOSi
MW: 199.25 g/mol
Boiling Pt: 130 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00000411
CAS Number: 24589-78-4
EINECS: 246-331-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a0715dae01d88ca2eb3929384f2f0c16831c688&vid=9a0715dae01d88ca2eb3929384f2f0c16831c688&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88a9d1582b58668d76c99be4fc9b4e17da5be019&vid=88a9d1582b58668d76c99be4fc9b4e17da5be019&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR