1-Methyl-2-pyridone

Warning

Synonyms: 1-Methylpyridin-2-ol, N-Methylpyridin-2-ol, 1-Methylpyridin-2(1H)-one, N-Methyl-2-pyridone

M78259-100G M78259-25G
220198-872EA 488.92 USD
220198-872 220198-873
1-Methyl-2-pyridone
1-Methyl-2-pyridone
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 33 °C
Density: 1.1321 g/cm³ (18 °C)
MDL Number: MFCD00006520
CAS Number: 694-85-9
EINECS: 211-777-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR