2-Mesitylenesulfonyl chloride ≥99%

Danger

Synonyms: 2,4,6-Trimethylbenzenesulphonyl chloride, Mesitylene-2-sulphonyl chloride

M7707-25G M7707-5G M7707-100G
220197-705EA 94.45 USD
220197-705 220197-706 220197-704
2-Mesitylenesulfonyl chloride ≥99%
2-Mesitylenesulphonyl chloride
Formula: C₉H₁₁ClO₂S
MW: 218.7 g/mol
Boiling Pt: 150 °C (20 -rr)
Melting Pt: 53 - 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00007434
CAS Number: 773-64-8
EINECS: 212-257-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR