Manganese(II) sulfate monohydrate

Warning

Synonyms: Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate

M7634-1KG M7634-100G M7634-500G
220198-821EA 72 USD
220198-821 220198-820 220198-822
Manganese(II) sulfate monohydrate
Manganese(II) sulphate monohydrate
Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700 °C
Density: 2.95 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 13,05763

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR