2-Methylpyrazine

M75608-100G M75608-25G
220198-802EA 58.15 USD
220198-802 220198-803
2-Methylpyrazine
2-Methylpyrazine
Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: -30 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006142
CAS Number: 109-08-0
EINECS: 203-645-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR