4-Methylpiperidine

M73206-5ML M73206-100ML M73206-500ML
220198-733EA 31.23 USD
220198-733 220198-731 220198-732
4-Methylpiperidine
4-Methylpiperidine
Formula: C₆H₁₃N
Boiling Pt: 124 °C (1013 hPa)
Melting Pt: -4 °C
Density: 0.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006005
CAS Number: 626-58-4
EINECS: 210-954-1
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR