3-Methylpiperidine

M73001-100G M73001-5G
220198-727EA 51.69 USD
220198-727 220198-728
3-Methylpiperidine
3-Methylpiperidine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 125 °C (1013 hPa)
Melting Pt: -24 °C
Density: 0.8635 g/cm³ (0 °C)
MDL Number: MFCD00005994
CAS Number: 626-56-2
EINECS: 210-953-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR