1-Methylpiperidine

M72609-500ML M72609-100ML
220198-714EA 149.69 USD
220198-714 220198-713
1-Methylpiperidine
1-Methylpiperidine
Formula: C₆H₁₃N
MW: 99.18 g/mol
Boiling Pt: 106.5 °C (1013 hPa)
Melting Pt: –69 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 3 °C
Storage Temperature: Freezer
MDL Number: MFCD00006491
CAS Number: 626-67-5
EINECS: 210-959-9
UN: 2399
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR