Methyl cellulose, 15 cP (2% H₂O)

Synonyms: Cellulose methyl ether

M7140-1KG M7140-500G M7140-250G M7140-100G
220198-157EA 423.38 USD
220198-157 220198-159 220198-158 220198-156
Methyl cellulose, 15 cP (2% H₂O)
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR