2,4,6-Trimethylbenzaldehyde ≥98%

Synonyms: Mesitaldehyde

M6808-10G M6808-50G
220198-567EA 161.54 USD
220198-567 220198-568
2,4,6-Trimethylbenzaldehyde ≥98%
2,4,6-Trimethylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 14 °C
Density: 1.0987 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003341
CAS Number: 487-68-3
EINECS: 207-662-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR