2-Methyl-2-pentene ≥98%

Synonyms: 1,1-Dimethyl-1-butene

M67303-5G M67303-25G
220196-855EA 178.77 USD
220196-855 220198-548
2-Methyl-2-pentene ≥98%
2-Methyl-2-pentene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 67 °C (1013 hPa)
Melting Pt: -135 °C
Density: 0.6904 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009382
CAS Number: 625-27-4
EINECS: 210-883-6
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR