3-Methyl-2-pentanone ≥99%

M67001-5G M67001-25G
220198-547EA 93.37 USD
220198-547 220198-546
3-Methyl-2-pentanone ≥99%
3-Methyl-2-pentanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 117 °C (1013 hPa)
Density: 0.815 g/cm³ (20 °C)
MDL Number: MFCD00009336
CAS Number: 565-61-7
EINECS: 209-282-1
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR