Methyl cellulose, 25 mPa.S (2% H₂O), Powder

Synonyms: Cellulose methyl ether

M6385-250G M6385-500G M6385-100G
220198-382EA 129.23 USD
220198-382 220198-383 220198-381
Methyl cellulose, 25 mPa.S (2% H₂O), Powder
Methyl cellulose

1 mPa.s = 1cP

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR