Methyl nicotinate

Synonyms: 3-Picolinic acid methyl ester, 3-(Methoxycarbonyl)pyridine, Pyridine-3-carboxylic acid methyl ester, Nicotinic acid methyl ester, 3-Pyridinecarboxylic acid methyl ester, Methyl 3-pyridinecarboxylate

M59203-100G M59203-500G M59203-5G
220198-271EA 47.38 USD
220198-271 220198-272 220198-273
Methyl nicotinate
Methyl nicotinate
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: 39 °C
Density: 1.0048 g/cm³ (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00006388
CAS Number: 93-60-7
EINECS: 202-261-8
Merck Index: 12,06174

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR