3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)

Warning

Synonyms: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide

M5655-500MG M5655-1G M5655-100MG
220196-114EA 128 USD
220196-114 220198-196 220198-195
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formula: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR