Methyl methacrylate ≥99%

Danger

Synonyms: Methacrylic acid methyl ester

M55909-25ML M55909-2L M55909-17L M55909-1L M55909-500ML
220197-688EA 45.89 USD
220197-688 220198-178 220197-686 220197-687 220198-179
Methyl methacrylate ≥99%
Methyl methacrylate
Formula: CH₂=C(CH₃)COOCH₃
MW: 100.12 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: –48 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008587
CAS Number: 80-62-6
EINECS: 201-297-1
UN: 1247
ADR: 3,II
REACH: 01-2119452498-28
Merck Index: 12,06005

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR