4-(Methylthio)aniline ≥98%

Synonyms: 4-(Methylmercapto)aniline

M54503-25G M54503-5G
220198-111EA 93.26 USD
220198-111 220198-112
4-(Methylthio)aniline ≥98%
4-(Methylthio)aniline
Formula: C₇H₉NS
MW: 139.22 g/mol
Boiling Pt: 74 °C (0.1 -rr)
Melting Pt: 156 - 158 °C
Density: 1.1379 g/cm³ (20 °C)
MDL Number: MFCD00007889
CAS Number: 104-96-1
EINECS: 203-256-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR