Methylmalonic acid ≥99%

Synonyms: 2-Methylmalonic acid

M54058-5G M54058-25G
220198-106EA 68.06 USD
220198-106 220198-105
Methylmalonic acid ≥99%
Methylmalonic acid
Formula: C₄H₆O₄
MW: 118.09 g/mol
MDL Number: MFCD00002656
CAS Number: 516-05-2
EINECS: 208-219-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR