3-Methylindole

Synonyms: Skatol, 3-Methyl-1H-indole, β-Methylindole, Skatole

M51458-100G M51458-5G M51458-25G
220198-034EA 372.62 USD
220198-034 220198-036 220198-035
3-Methylindole
3-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 95…97 °C
Density: 1.003 g/cm³ (99.6 °C)
MDL Number: MFCD00005627
CAS Number: 83-34-1
EINECS: 201-471-7
Merck Index: 12,08700

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR