2-Methylimidazole

Synonyms: 2-Methyl-1H-imidazole

M50850-500G M50850-100G
220198-026EA 286.46 USD
220198-026 220198-025
2-Methylimidazole
2-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 130…133 °C
Density: 0.99 g/cm³ (25 °C)
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR