1-Methylimidazole (NMI)

Danger

M50834-100G M50834-500G M50834-3KG
220198-022EA 45.12 USD
220198-022 220198-024 220198-023
1-Methylimidazole (NMI)
1-Methylimidazole
Formula: C₄H₆N₂
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
EINECS: 210-484-7
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR