Methylhydrazine ≥98%

M50001-100G M50001-25G
220196-105EA 268.15 USD
220196-105 220196-106
Methylhydrazine ≥98%
Methylhydrazine
Formula: CH₆N₂
MW: 46.07 g/mol
CAS Number: 60-34-4
EINECS: 200-471-4
UN: 1244
ADR: 6.1,I

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR