6-Methyl-5-hepten-2-one ≥99%

Synonyms: 2-Methyl-2-hepten-6-one, Sulcatone

M48805-100ML M48805-5ML
220197-978EA 38.66 USD
220197-978 220197-979
6-Methyl-5-hepten-2-one ≥99%
6-Methyl-5-hepten-2-one
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –67.1 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00008905
CAS Number: 110-93-0
EINECS: 203-816-7
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR