1,3,3-Trimethyl-2-methyleneindoline

M46209-100G M46209-5G
220197-936EA 188.46 USD
220197-936 220197-937
1,3,3-Trimethyl-2-methyleneindoline
1,3,3-Trimethyl-2-methyleneindoline
Formula: C₁₂H₁₅N
MW: 173.26 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.986 g/cm³ (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00005813
CAS Number: 118-12-7
EINECS: 204-235-1
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR