Mercury(II) thiocyanate

Danger

Synonyms: Mercury dithiocyanate, Mercuric thiocyanate, Mercury (II) rhodanide

M4128-100G M4128-25G
220197-775EA 144.69 USD
220197-775 220197-777
Mercury(II) thiocyanate
Mercury(II) thiocyanate
Formula: C₂HgN₂S₂
MW: 316.76 g/mol
Melting Pt: –38.9 °C
Density: 4 g/cm³ (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00011039
CAS Number: 592-85-8
EINECS: 209-773-0
UN: 1646
ADR: 6.1,II
Merck Index: 13,05917

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR