4-Methylpyrocatechol ≥95%

Synonyms: 4-Methylbenzene-1,2-diol, 4-Methylcatechol

M34200-500G M34200-100G M34200-5G
220197-615EA 323.08 USD
220197-615 220197-614 220197-616
4-Methylpyrocatechol ≥95%
4-Methylpyrocatechol
Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00002205
CAS Number: 452-86-8
EINECS: 207-214-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR