2-Mercaptobenzothiazole

Warning

Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT

M3302-1KG M3302-5G M3302-100G
220197-591EA 70.97 USD
220197-591 220197-592 220197-590
2-Mercaptobenzothiazole
2-Mercaptobenzothiazole
Formula: C₇H₅NS₂
MW: 167.26 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 177…182 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Merck Index: 13,05893

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR