2-Methyl-1-butene-3-yne

Synonyms: 2-Methylbut-1-en-3-yne

M32801-25G M32801-5G
220197-583EA 328.85 USD
220197-583 220197-584
2-Methyl-1-butene-3-yne
2-Methyl-1-butene-3-yne
Formula: C₅H₆
MW: 66.1 g/mol
Boiling Pt: 32 °C (1013 hPa)
Melting Pt: –113 °C
Density: 1.001 g/cm³ (25 °C)
MDL Number: MFCD00008599
CAS Number: 78-80-8
EINECS: 201-144-9
UN: 3295
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR