2-Methyl-2-butene ≥99%

Danger

Synonyms: Trimethylethylene, Isoamylene, Isopentene

M32704-100ML M32704-500ML
220197-579EA 147.54 USD
220197-579 220197-580
2-Methyl-2-butene ≥99%
2-Methyl-2-butene
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 39 °C (1013 hPa)
Melting Pt: -124 °C
Density: 0.6623 g/cm³ (20 °C)
Flash Pt: -45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009276
CAS Number: 513-35-9
EINECS: 208-156-3
UN: 2460
ADR: 3,II
Merck Index: 12,00649

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR