2-Mercaptobenzimidazole

Synonyms: Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol

M3205-100G M3205-500G M3205-5G
220197-560EA 50.83 USD
220197-560 220197-561 220197-562
2-Mercaptobenzimidazole
2-Mercaptobenzimidazole
Formula: C₇H₆N₂S
MW: 150.2 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 154 °C
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00466107
CAS Number: 583-39-1
EINECS: 209-502-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR