2-Mercaptoethanol for electrophoresis

Danger

M3148-100ML M3148-250ML M3148-25ML
220197-530EA 91.03 USD
220197-530 220197-531 220197-532
2-Mercaptoethanol for electrophoresis
2-Mercaptoethanol
Formula: HSCH₂CH₂OH
Boiling Pt: 157 °C (1013 hPa)
Melting Pt: –100 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00004890
CAS Number: 60-24-2
EINECS: 200-464-6
UN: 2966
ADR: 6.1,II
Merck Index: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc427209b84d0343a5ae47c435ddfc53df4013d2&vid=7d70fd087d948d54e7a946d3b2310e2d820d2396&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR